Assay Details
| Gene Symbol: Ppp1r3g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497486.2 | XP_003497534.1 | 2 | 1719 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppp1r3g| Gene Symbol: Ppp1r3g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497486.2 | XP_003497534.1 | 2 | 1719 | 99 | |
Set Membership: |
|