Assay Details
| Gene Symbol: Oxtr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498097.2 | XP_003498145.2 | 2 | 1045 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Oxtr| Gene Symbol: Oxtr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498097.2 | XP_003498145.2 | 2 | 1045 | 77 | |
Set Membership: |
|