Assay Details
| Gene Symbol: Nfil3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003508185.2 | XP_003508233.1 | 2 | 1219 | 90 | |
| XM_007648808.1 | XP_007646998.1 | 2 | 1219 | 90 | ||
More Information
Set Membership: |
|

Nfil3
Genomic Map