Assay Details
| Gene Symbol: PPP4C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039937.1 | XP_017895426.1 | 8 - 9 | 971 | 54 | |
| GenBank mRNA | JT762484.1 | - | 1 - 2 | 389 | 54 | |
More Information
Set Membership: |
|

PPP4C
Genomic Map