Assay Details
| Gene Symbol: PAIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065512.1 | XP_017921001.1 | 4 - 5 | 994 | 70 | |
| XM_018065513.1 | XP_017921002.1 | 4 - 5 | 748 | 70 | ||
| XM_018065514.1 | XP_017921003.1 | 4 - 5 | 598 | 70 | ||
| GenBank mRNA | JT669100.1 | - | 4 - 5 | 542 | 70 | |
More Information
Set Membership: |
|

PAIP1
Genomic Map