Assay Details
| Gene Symbol: PCBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018047662.1 | XP_017903151.1 | 3 - 4 | 282 | 66 | |
| XM_018047663.1 | XP_017903152.1 | 3 - 4 | 282 | 66 | ||
| XM_018047664.1 | XP_017903153.1 | 3 - 4 | 438 | 66 | ||
| XM_018047665.1 | XP_017903154.1 | 3 - 4 | 443 | 66 | ||
| XM_018047666.1 | XP_017903155.1 | 3 - 4 | 282 | 66 | ||
| XM_018047667.1 | XP_017903156.1 | 3 - 4 | 282 | 66 | ||
| XM_018047668.1 | XP_017903157.1 | 3 - 4 | 281 | 66 | ||
| XM_018047669.1 | XP_017903158.1 | 3 - 4 | 281 | 66 | ||
| XM_018047670.1 | XP_017903159.1 | 3 - 4 | 282 | 66 | ||
| XM_018047671.1 | XP_017903160.1 | 3 - 4 | 443 | 66 | ||
| XM_018047672.1 | XP_017903161.1 | 3 - 4 | 282 | 66 | ||
| XM_018047673.1 | XP_017903162.1 | 3 - 4 | 282 | 66 | ||
| XM_018047674.1 | XP_017903163.1 | 3 - 4 | 282 | 66 | ||
| XM_018047675.1 | XP_017903164.1 | 3 - 4 | 443 | 66 | ||
| XM_018047676.1 | XP_017903165.1 | 3 - 4 | 282 | 66 | ||
| GenBank mRNA | JO581242.1 | - | 6 - 7 | 639 | 66 | |
| JO581243.1 | - | 6 - 7 | 651 | 66 | ||
More Information
Set Membership: |
|

PCBP2
Genomic Map