Assay Details
| Gene Symbol: COP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060629.1 | XP_017916118.1 | 13 - 14 | 1641 | 95 | |
| XM_018060630.1 | XP_017916119.1 | 13 - 14 | 1629 | 95 | ||
| XM_018060631.1 | XP_017916120.1 | 12 - 13 | 1572 | 95 | ||
More Information
Set Membership: |
|

COP1
Genomic Map