Assay Details
| Gene Symbol: SP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018047337.1 | XP_017902826.1 | 2 - 3 | 332 | 111 | |
| XM_018047338.1 | XP_017902827.1 | 2 - 3 | 209 | 111 | ||
More Information
Set Membership: |
|

SP4
Genomic Map