Assay Details
| Gene Symbol: CTPS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018042505.1 | XP_017897994.1 | 11 - 12 | 1341 | 90 | |
| XM_018042518.1 | XP_017898007.1 | 11 - 12 | 2164 | 90 | ||
| XM_018042523.1 | XP_017898012.1 | 8 - 9 | 844 | 90 | ||
| GenBank mRNA | KA323966.1 | - | 11 - 12 | 1270 | 90 | |
More Information
Set Membership: |
|

CTPS1
Genomic Map