Assay Details
| Gene Symbol: DAZL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001314241.1 | NP_001301170.1 | 4 - 5 | 333 | 71 | |
| GenBank mRNA | JX183813.1 | - | 3 - 4 | 212 | 71 | |
| JX273761.1 | - | 4 - 5 | 333 | 71 | ||
| KC189823.1 | - | 3 - 4 | 234 | 71 | ||
More Information
Set Membership: |
|

DAZL
Genomic Map