Assay Details
| Gene Symbol: SLC22A17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054599.1 | XP_017910088.1 | 5 - 6 | 2860 | 60 | |
| XM_018054600.1 | XP_017910089.1 | 5 - 6 | 2858 | 60 | ||
More Information
Set Membership: |
|

SLC22A17
Genomic Map