Assay Details
| Gene Symbol: RXRB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018038584.1 | XP_017894073.1 | 2 - 3 | 694 | 55 | |
| XM_018038585.1 | XP_017894074.1 | 2 - 3 | 694 | 55 | ||
| GenBank mRNA | JO454038.2 | - | 2 - 3 | 407 | 55 | |
More Information
Set Membership: |
|

RXRB
Genomic Map