Assay Details
Gene Symbol: TENT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005685076.3 | XP_005685133.2 | 7 - 8 | 1083 | 67 | |
XM_005685077.3 | XP_005685134.2 | 5 - 6 | 903 | 67 | ||
XM_005685078.3 | XP_005685135.2 | 6 - 7 | 1012 | 67 | ||
XM_005685080.3 | XP_005685137.2 | 7 - 8 | 824 | 67 | ||
XM_005685081.3 | XP_005685138.2 | 5 - 6 | 613 | 67 | ||
XM_005685083.3 | XP_005685140.2 | 5 - 6 | 892 | 67 | ||
XM_005685085.3 | XP_005685142.2 | 5 - 6 | 903 | 67 | ||
XM_013966728.2 | XP_013822182.2 | 6 - 7 | 1701 | 67 | ||
XM_013966730.2 | XP_013822184.2 | 6 - 7 | 1451 | 67 | ||
XM_013966733.2 | XP_013822187.2 | 6 - 7 | 1701 | 67 | ||
XM_018053670.1 | XP_017909159.1 | 6 - 7 | 943 | 67 | ||
XM_018053671.1 | XP_017909160.1 | 7 - 8 | 1078 | 67 | ||
XM_018053672.1 | XP_017909161.1 | 6 - 7 | 753 | 67 | ||
GenBank mRNA | JO578495.1 | - | 11 - 12 | 1173 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
