Assay Details
| Gene Symbol: LOC102181832 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040400.1 | XP_017895889.1 | 12 - 13 | 1718 | 69 | |
| GenBank mRNA | JO360672.2 | - | 4 - 5 | 961 | 69 | |
More Information
Set Membership: |
|

LOC102181832
Genomic Map