Assay Details
| Gene Symbol: YTHDF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057048.1 | XP_017912537.1 | 3 - 4 | 640 | 89 | |
| GenBank mRNA | JO635385.2 | - | 3 - 4 | 318 | 89 | |
More Information
Set Membership: |
|

YTHDF2
Genomic Map