Assay Details
| Gene Symbol: SLC40A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065526.1 | XP_017921015.1 | 7 - 8 | 1714 | 73 | |
| GenBank mRNA | KA321854.1 | - | 8 - 9 | 1718 | 73 | |
More Information
Set Membership: |
|

SLC40A1
Genomic Map