Assay Details
| Gene Symbol: PAK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018046304.1 | XP_017901793.1 | 3 - 4 | 535 | 66 | |
| GenBank mRNA | JO339808.2 | - | 10 - 11 | 898 | 66 | |
| JO367180.2 | - | 5 - 6 | 478 | 66 | ||
| JO388345.2 | - | 3 - 4 | 386 | 66 | ||
| JO409974.2 | - | 3 - 4 | 504 | 66 | ||
| JO431167.2 | - | 12 - 13 | 1316 | 66 | ||
| JO431899.2 | - | 12 - 13 | 1316 | 66 | ||
| JO499289.2 | - | 3 - 4 | 420 | 66 | ||
| JO613591.2 | - | 12 - 13 | 2263 | 66 | ||
| JO633727.2 | - | 2 - 3 | 164 | 66 | ||
| JT762738.1 | - | 12 - 13 | 1242 | 66 | ||
| KA315896.1 | - | 3 - 4 | 362 | 66 | ||
More Information
Set Membership: |
|

PAK2
Genomic Map