Assay Details
| Gene Symbol: MAP1LC3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061829.1 | XP_017917318.1 | 2 - 3 | 238 | 69 | |
| GenBank mRNA | JO528456.2 | - | 1 - 2 | 62 | 69 | |
| JO634892.2 | - | 2 - 3 | 245 | 69 | ||
More Information
Set Membership: |
|

MAP1LC3B
Genomic Map