Assay Details
| Gene Symbol: HFE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018038509.1 | XP_017893998.1 | 4 - 5 | 1079 | 55 | |
| GenBank mRNA | JO594282.2 | - | 2 - 3 | 1164 | 55 | |
More Information
Set Membership: |
|

HFE
Genomic Map