Assay Details
| Gene Symbol: ZSCAN21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039892.1 | XP_017895381.1 | 3 - 4 | 769 | 77 | |
| XM_018039893.1 | XP_017895382.1 | 3 - 4 | 836 | 77 | ||
| XM_018039894.1 | XP_017895383.1 | 3 - 4 | 702 | 77 | ||
| GenBank mRNA | JO430440.2 | - | 4 - 5 | 782 | 77 | |
More Information
Set Membership: |
|

ZSCAN21
Genomic Map