Assay Details
| Gene Symbol: GOLGA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018042031.1 | XP_017897520.1 | 4 - 5 | 496 | 71 | |
| XM_018042032.1 | XP_017897521.1 | 4 - 5 | 480 | 71 | ||
| GenBank mRNA | JO430780.2 | - | 2 - 3 | 1428 | 71 | |
| JO431279.2 | - | 2 - 3 | 1428 | 71 | ||
| JO503509.2 | - | 3 - 4 | 540 | 71 | ||
More Information
Set Membership: |
|

GOLGA7
Genomic Map