Assay Details
| Gene Symbol: GCG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005676044.3 | XP_005676101.1 | 4 - 5 | 491 | 74 | |
| XM_005676045.3 | XP_005676102.1 | 4 - 5 | 491 | 74 | ||
More Information
Set Membership: |
|

GCG
Genomic Map