Assay Details
| Gene Symbol: OMD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005684164.3 | XP_005684221.1 | 2 - 3 | 1087 | 118 | |
| GenBank mRNA | JT660890.1 | - | 1 - 2 | 408 | 118 | |
More Information
Set Membership: |
|

OMD
Genomic Map