Assay Details
| Gene Symbol: CLTA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018052167.1 | XP_017907656.1 | 3 - 4 | 684 | 58 | |
| XM_018052168.1 | XP_017907657.1 | 3 - 4 | 684 | 58 | ||
| XM_018052169.1 | XP_017907658.1 | 3 - 4 | 684 | 58 | ||
| XM_018052171.1 | XP_017907660.1 | 3 - 4 | 684 | 58 | ||
More Information
Set Membership: |
|

CLTA
Genomic Map