Assay Details
Gene Symbol: PPM1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018054085.1 | XP_017909574.1 | 3 - 4 | 1389 | 95 | |
GenBank mRNA | JT755470.1 | - | 1291 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
