Assay Details
| Gene Symbol: ACADVL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005693483.3 | XP_005693540.1 | 10 - 11 | 1177 | 63 | |
| XM_018064553.1 | XP_017920042.1 | 10 - 11 | 1176 | 63 | ||
| GenBank mRNA | JO621268.2 | - | 10 - 11 | 1207 | 63 | |
More Information
Set Membership: |
|

ACADVL
Genomic Map