Assay Details
| Gene Symbol: LOC102179132 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064502.1 | XP_017919991.1 | 3 - 4 | 1884 | 131 | |
| XM_018064503.1 | XP_017919992.1 | 4 - 5 | 1906 | 131 | ||
| GenBank mRNA | JO332083.2 | - | 7 - 8 | 1162 | 131 | |
| JO548077.2 | - | 4 - 5 | 571 | 131 | ||
| KA309795.1 | - | 4 - 5 | 695 | 131 | ||
More Information
Set Membership: |
|

LOC102179132
Genomic Map