Assay Details
| Gene Symbol: ENTPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005698264.3 | XP_005698321.1 | 2 - 3 | 207 | 67 | |
| XM_013975163.2 | XP_013830617.1 | 2 - 3 | 326 | 67 | ||
| GenBank mRNA | JO338693.2 | - | 4 - 5 | 1076 | 67 | |
| JO585652.2 | - | 2 - 3 | 357 | 67 | ||
More Information
Set Membership: |
|

ENTPD1
Genomic Map