Assay Details
Gene Symbol: LOC108633300 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018044709.1 | XP_017900198.1 | 2 - 3 | 455 | 86 | |
GenBank mRNA | JO641536.2 | - | 384 | 86 |
Gene Symbol: GABRA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018044329.1 | XP_017899818.1 | 142 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
