Assay Details
Gene Symbol: SLC2A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001314223.1 | NP_001301152.1 | 4 - 5 | 624 | 62 | |
GenBank mRNA | JO382759.2 | - | 1 - 2 | 134 | 62 | |
JQ343217.1 | - | 4 - 5 | 624 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
