Assay Details
| Gene Symbol: CSNK1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683152.3 | XP_005683209.1 | 8 - 9 | 1413 | 80 | |
| XM_005683153.3 | XP_005683210.1 | 8 - 9 | 1412 | 80 | ||
| XM_005683154.3 | XP_005683211.1 | 7 - 8 | 1329 | 80 | ||
| XM_005683156.3 | XP_005683213.1 | 7 - 8 | 1329 | 80 | ||
| XM_018050173.1 | XP_017905662.1 | 8 - 9 | 1412 | 80 | ||
| XM_018050174.1 | XP_017905663.1 | 8 - 9 | 1412 | 80 | ||
| XM_018050175.1 | XP_017905664.1 | 7 - 8 | 1328 | 80 | ||
| XM_018050176.1 | XP_017905665.1 | 7 - 8 | 1328 | 80 | ||
| XM_018050177.1 | XP_017905666.1 | 7 - 8 | 1328 | 80 | ||
| GenBank mRNA | KA314985.1 | - | 3 - 4 | 1202 | 80 | |
More Information
Set Membership: |
|

CSNK1A1
Genomic Map