Assay Details
| Gene Symbol: LOC102183306 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018049273.1 | XP_017904762.1 | 3 - 4 | 406 | 62 | |
| GenBank mRNA | JO404007.2 | - | 3 - 4 | 393 | 62 | |
| JO579992.1 | - | 3 - 4 | 463 | 62 | ||
More Information
Set Membership: |
|

LOC102183306
Genomic Map