Assay Details
| Gene Symbol: LOC102187632 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005676339.3 | XP_005676396.1 | 33 - 34 | 4133 | 65 | |
| GenBank mRNA | JO422252.2 | - | 1548 | 65 | ||
More Information
Set Membership: |
|

LOC102187632
Genomic Map