Assay Details
| Gene Symbol: TRIB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005686991.3 | XP_005687048.1 | 1 - 2 | 1700 | 73 | |
| XM_018055626.1 | XP_017911115.1 | 1 - 2 | 1688 | 73 | ||
| GenBank mRNA | JO348988.2 | - | 1 - 2 | 824 | 73 | |
| JO579458.1 | - | 2 - 3 | 1488 | 73 | ||
More Information
Set Membership: |
|

TRIB2
Genomic Map