Assay Details
| Gene Symbol: PTGER3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018045482.1 | XP_017900971.1 | 1 - 2 | 1204 | 105 | |
| XM_018045484.1 | XP_017900973.1 | 1 - 2 | 1204 | 105 | ||
| XM_018045488.1 | XP_017900977.1 | 1 - 2 | 1204 | 105 | ||
| GenBank mRNA | JO433953.2 | - | 3 - 4 | 349 | 105 | |
| JO624190.2 | - | 1 - 2 | 132 | 105 | ||
More Information
Set Membership: |
|

PTGER3
Genomic Map