Assay Details
| Gene Symbol: NME1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005693641.3 | XP_005693698.1 | 3 - 4 | 426 | 76 | |
| XM_005693642.3 | XP_005693699.1 | 3 - 4 | 481 | 76 | ||
| XM_005693645.3 | XP_005693702.1 | 3 - 4 | 501 | 76 | ||
| XM_005693646.3 | XP_005693703.1 | 3 - 4 | 509 | 76 | ||
| GenBank mRNA | JO421632.2 | - | 3 - 4 | 396 | 76 | |
| JO421633.2 | - | 3 - 4 | 464 | 76 | ||
More Information
Set Membership: |
|

NME1
Genomic Map