Assay Details
| Gene Symbol: CCNG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050079.1 | XP_017905568.1 | 4 - 5 | 828 | 72 | |
| XM_018050080.1 | XP_017905569.1 | 4 - 5 | 826 | 72 | ||
| GenBank mRNA | JO366531.2 | - | 3 - 4 | 1532 | 72 | |
| JO428518.1 | - | 3 - 4 | 1599 | 72 | ||
More Information
Set Membership: |
|

CCNG1
Genomic Map