Assay Details
Gene Symbol: MTG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018057590.1 | XP_017913079.1 | 5 - 6 | 528 | 72 | |
XM_018057591.1 | XP_017913080.1 | 5 - 6 | 1089 | 72 | ||
XM_018057592.1 | XP_017913081.1 | 4 - 5 | 519 | 72 | ||
GenBank mRNA | KA312139.1 | - | 3 - 4 | 479 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
