Assay Details
| Gene Symbol: PRICKLE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005700749.3 | XP_005700806.2 | 2 - 3 | 244 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRICKLE3| Gene Symbol: PRICKLE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005700749.3 | XP_005700806.2 | 2 - 3 | 244 | 61 | |
Set Membership: |
|