Assay Details
| Gene Symbol: NFYA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039157.1 | XP_017894646.1 | 5 - 6 | 684 | 60 | |
| XM_018039158.1 | XP_017894647.1 | 5 - 6 | 681 | 60 | ||
| XM_018039159.1 | XP_017894648.1 | 5 - 6 | 681 | 60 | ||
| XM_018039160.1 | XP_017894649.1 | 5 - 6 | 684 | 60 | ||
| XM_018039161.1 | XP_017894650.1 | 4 - 5 | 597 | 60 | ||
| XM_018039162.1 | XP_017894651.1 | 4 - 5 | 597 | 60 | ||
| GenBank mRNA | KA323891.1 | - | 4 - 5 | 538 | 60 | |
More Information
Set Membership: |
|

NFYA
Genomic Map