Assay Details
| Gene Symbol: HNF4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005688574.3 | XP_005688631.1 | 6 - 7 | 899 | 60 | |
| XM_005688575.3 | XP_005688632.1 | 6 - 7 | 899 | 60 | ||
| XM_013968895.2 | XP_013824349.1 | 6 - 7 | 1101 | 60 | ||
| XM_013968896.2 | XP_013824350.2 | 6 - 7 | 899 | 60 | ||
| GenBank mRNA | JO420888.2 | - | 6 - 7 | 932 | 60 | |
| JO420889.2 | - | 6 - 7 | 932 | 60 | ||
More Information
Set Membership: |
|

HNF4A
Genomic Map