Assay Details
Gene Symbol: BSCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018042981.1 | XP_017898470.1 | 6 - 7 | 971 | 57 | |
XM_018042982.1 | XP_017898471.1 | 6 - 7 | 937 | 57 | ||
XM_018042983.1 | XP_017898472.1 | 5 - 6 | 1226 | 57 | ||
XM_018042984.1 | XP_017898473.1 | 5 - 6 | 862 | 57 | ||
XM_018042985.1 | XP_017898474.1 | 5 - 6 | 936 | 57 | ||
XM_018042986.1 | XP_017898475.1 | 5 - 6 | 936 | 57 | ||
XM_018042987.1 | XP_017898476.1 | 4 - 5 | 631 | 57 | ||
XM_018042988.1 | XP_017898477.1 | 4 - 5 | 670 | 57 | ||
GenBank mRNA | JT763332.1 | - | 6 - 7 | 2450 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
