Assay Details
| Gene Symbol: C3H1orf43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677439.3 | XP_005677496.2 | 1 - 2 | 447 | 90 | |
| XM_018046188.1 | XP_017901677.1 | 1 - 2 | 447 | 90 | ||
| GenBank mRNA | JO373438.2 | - | 1429 | 90 | ||
| JO499345.2 | - | 6 - 7 | 1438 | 90 | ||
| Gene Symbol: LOC102177858 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040521.1 | XP_017896010.1 | 1 | 246 | 90 | |
More Information
Set Membership: |
|

C3H1orf43
Genomic Map