Assay Details
| Gene Symbol: SLC46A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013966271.2 | XP_013821725.2 | 1 - 2 | 1926 | 76 | |
| GenBank mRNA | KA328991.1 | - | 3 - 4 | 1032 | 76 | |
More Information
Set Membership: |
|

SLC46A2
Genomic Map