Assay Details
| Gene Symbol: TMEM54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018058059.1 | XP_017913548.1 | 4 - 5 | 633 | 59 | |
| XM_018058060.1 | XP_017913549.1 | 3 - 4 | 573 | 59 | ||
| XM_018058061.1 | XP_017913550.1 | 4 - 5 | 633 | 59 | ||
| XM_018058063.1 | XP_017913552.1 | 3 - 4 | 573 | 59 | ||
| GenBank mRNA | KA348401.1 | - | 2 - 3 | 917 | 59 | |
More Information
Set Membership: |
|

TMEM54
Genomic Map