Assay Details
Gene Symbol: LOC102179319 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013962506.2 | XP_013817960.1 | 7 - 8 | 1075 | 107 | |
GenBank mRNA | JO382243.2 | - | 2 - 3 | 538 | 107 | |
JO543593.2 | - | 1 - 2 | 89 | 107 | ||
JO632563.2 | - | 2 - 3 | 659 | 107 | ||
KA346920.1 | - | 1 - 2 | 89 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() |
