Assay Details
| Gene Symbol: SNRPB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057446.1 | XP_017912935.1 | 2 - 3 | 286 | 71 | |
| XM_018057447.1 | XP_017912936.1 | 2 - 3 | 286 | 71 | ||
| GenBank mRNA | JO581975.1 | - | 2 - 3 | 337 | 71 | |
| JO581976.1 | - | 2 - 3 | 337 | 71 | ||
More Information
Set Membership: |
|

SNRPB
Genomic Map