Assay Details
| Gene Symbol: TSPAN18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018059204.1 | XP_017914693.1 | 9 - 10 | 1218 | 57 | |
| XM_018059205.1 | XP_017914694.1 | 8 - 9 | 1077 | 57 | ||
| XM_018059206.1 | XP_017914695.1 | 9 - 10 | 1207 | 57 | ||
| XM_018059207.1 | XP_017914696.1 | 8 - 9 | 1066 | 57 | ||
| GenBank mRNA | KA348166.1 | - | 2 - 3 | 1064 | 57 | |
More Information
Set Membership: |
|

TSPAN18
Genomic Map