Assay Details
| Gene Symbol: AIF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696600.3 | XP_005696657.1 | 2 - 3 | 261 | 70 | |
| XM_005696601.3 | XP_005696658.1 | 2 - 3 | 284 | 70 | ||
| GenBank mRNA | JI861623.1 | - | 2 - 3 | 86 | 70 | |
| KX068697.1 | - | 2 - 3 | 90 | 70 | ||
More Information
Set Membership: |
|

AIF1
Genomic Map