Assay Details
| Gene Symbol: VEGFB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018042785.1 | XP_017898274.1 | 3 - 4 | 551 | 55 | |
| XM_018042786.1 | XP_017898275.1 | 3 - 4 | 551 | 55 | ||
| XM_018042787.1 | XP_017898276.1 | 3 - 4 | 551 | 55 | ||
| GenBank mRNA | JO393043.2 | - | 3 - 4 | 503 | 55 | |
More Information
Set Membership: |
|

VEGFB
Genomic Map